Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504468.2 | XP_003504516.1 | 3 - 4 | 525 | 65 | |
XM_007645008.1 | XP_007643198.1 | 3 - 4 | 448 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |