Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aph1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498637.2 | XP_003498685.1 | 1 - 2 | 328 | 85 | |
GenBank mRNA | JI874342.1 | - | 1 - 2 | 106 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |