Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3rf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503277.2 | XP_003503325.1 | 5 - 6 | 1583 | 84 | |
XM_007643880.1 | XP_007642070.1 | 5 - 6 | 1400 | 84 | ||
XM_007643881.1 | XP_007642071.1 | 5 - 6 | 1300 | 84 | ||
XM_007643882.1 | XP_007642072.1 | 5 - 6 | 1265 | 84 |