Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515101.2 | XP_003515149.2 | 2 - 3 | 142 | 73 | |
GenBank mRNA | JI888489.1 | - | 1 - 2 | 70 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |