Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651829.1 | XP_007650019.1 | 3 - 4 | 561 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Probe spans exons |