Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651829.1 | XP_007650019.1 | 6 - 7 | 1188 | 85 | |
XM_007651830.1 | XP_007650020.1 | 5 - 6 | 963 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Protease Probe spans exons |