Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497959.1 | XP_003498007.1 | 5 - 6 | 568 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |