Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc10a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498254.2 | XP_003498302.1 | 1 - 2 | 356 | 68 | |
XM_007625042.1 | XP_007623232.1 | 1 - 2 | 356 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Recommended Assay Probe spans exons |