Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdm12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513122.1 | XP_003513170.1 | 1 - 2 | 221 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |