Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007615481.1 | XP_007613671.1 | 2 - 3 | 503 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |