Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erp27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499208.2 | XP_003499256.1 | 6 - 7 | 774 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Probe spans exons |