Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cav2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500773.2 | XP_003500821.1 | 2 - 3 | 516 | 85 | |
GenBank mRNA | JI895493.1 | - | 2 - 3 | 408 | 85 | |
JP059184.1 | - | 2 - 3 | 408 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |