Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503907.2 | XP_003503955.1 | 7 - 8 | 950 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |