Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vstm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511705.2 | XP_003511753.1 | 2 - 3 | 241 | 96 | |
XM_007652240.1 | XP_007650430.1 | 2 - 3 | 241 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |