Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647513.1 | XP_007645703.1 | 4 - 5 | 521 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |