Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hfe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642819.1 | XP_007641009.1 | 5 - 6 | 1180 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Probe spans exons |