Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509544.1 | XP_003509592.1 | 1 - 2 | 403 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |