Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640567.1 | XP_007638757.1 | 5 - 6 | 511 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Transporter 3' Most Probe spans exons |