Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glyatl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510666.2 | XP_003510714.1 | 2 - 3 | 176 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |