Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515118.2 | XP_003515166.1 | 1 - 2 | 3934 | 62 | |
XM_007655127.1 | XP_007653317.1 | 1 - 2 | 3935 | 62 | ||
XM_007655128.1 | XP_007653318.1 | 1 - 2 | 3937 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |