Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Areg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007652867.1 | XP_007651057.1 | 5 - 6 | 1029 | 92 | |
GenBank mRNA | JI866423.1 | - | 1 - 2 | 44 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |