Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007624845.1 | XP_007623035.1 | 6 - 7 | 780 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel Probe spans exons |