Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007644286.1 | XP_007642476.1 | 5 - 6 | 2546 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Transporter 3' Most Detect Genomic DNA Probe spans exons |