Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501218.2 | XP_003501266.1 | 13 - 14 | 1667 | 89 | |
XM_007641906.1 | XP_007640096.1 | 13 - 14 | 1503 | 89 | ||
XM_007641907.1 | XP_007640097.1 | 12 - 13 | 1517 | 89 | ||
GenBank mRNA | JP061130.1 | - | 8 - 9 | 938 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |