Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501218.2 | XP_003501266.1 | 6 - 7 | 941 | 67 | |
XM_007641906.1 | XP_007640096.1 | 6 - 7 | 777 | 67 | ||
XM_007641907.1 | XP_007640097.1 | 6 - 7 | 941 | 67 | ||
GenBank mRNA | JP061130.1 | - | 1 - 2 | 212 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |