Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd79a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508884.1 | XP_003508932.1 | 4 - 5 | 508 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |