Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ninj1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007645879.1 | XP_007644069.1 | 2 - 3 | 504 | 104 | |
GenBank mRNA | JI862035.1 | - | 2 - 3 | 504 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |