Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500499.2 | XP_003500547.1 | 2 - 3 | 282 | 116 | |
GenBank mRNA | JI885879.1 | - | 2 - 3 | 93 | 116 | |
JP039303.1 | - | 2 - 3 | 93 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |