Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514588.2 | XP_003514636.1 | 1 - 2 | 113 | 87 | |
GenBank mRNA | JI870243.1 | - | 1 - 2 | 90 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |