Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007652818.1 | XP_007651008.1 | 2 - 3 | 279 | 92 | |
GenBank mRNA | JI895138.1 | - | 2 - 3 | 351 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |