Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007648738.1 | XP_007646928.1 | 6 - 7 | 1238 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Apoptosis Protease 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |