Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007633131.1 | XP_007631321.1 | 3 - 4 | 1544 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |