Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654245.1 | XP_007652435.1 | 1 - 2 | 135 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Protease 3' Most Detect Genomic DNA Probe spans exons |