Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007645440.1 | XP_007643630.1 | 6 - 7 | 784 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Probe spans exons |