Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641476.1 | XP_007639666.1 | 2 - 3 | 117 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |