Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495170.2 | XP_003495218.1 | 3 - 4 | 331 | 99 | |
GenBank mRNA | JP038227.1 | - | 3 - 4 | 289 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |