Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654974.1 | XP_007653164.1 | 4 - 5 | 1589 | 70 | |
XM_007654975.1 | XP_007653165.1 | 5 - 6 | 1462 | 70 | ||
XM_007654976.1 | XP_007653166.1 | 3 - 4 | 892 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |