Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654974.1 | XP_007653164.1 | 9 - 10 | 2231 | 83 | |
XM_007654975.1 | XP_007653165.1 | 10 - 11 | 2104 | 83 | ||
XM_007654976.1 | XP_007653166.1 | 8 - 9 | 1534 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |