Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007632011.1 | XP_007630201.1 | 3 - 4 | 206 | 87 | |
XM_007632020.1 | XP_007630210.1 | 3 - 4 | 343 | 87 | ||
GenBank mRNA | JP039729.1 | - | 2 - 3 | 278 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |