Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sord | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498024.2 | XP_003498072.1 | 8 - 9 | 1004 | 70 | |
GenBank mRNA | JI888278.1 | - | 8 - 9 | 994 | 70 | |
JP041914.1 | - | 8 - 9 | 994 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |