Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498255.2 | XP_003498303.1 | 6 - 7 | 556 | 69 | |
XM_007625054.1 | XP_007623244.1 | 6 - 7 | 556 | 69 | ||
GenBank mRNA | JI878755.1 | - | 1 - 2 | 73 | 69 | |
JP050528.1 | - | 1 - 2 | 70 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |