Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd30a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504266.2 | XP_003504314.1 | 26 - 27 | 4036 | 94 | |
XM_007644818.1 | XP_007643008.1 | 26 - 27 | 4051 | 94 | ||
XM_007644819.1 | XP_007643009.1 | 26 - 27 | 4051 | 94 | ||
GenBank mRNA | JI877755.1 | - | 3 - 4 | 1001 | 94 | |
JP048856.1 | - | 3 - 4 | 1000 | 94 |