Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd30a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504266.2 | XP_003504314.1 | 30 - 31 | 4669 | 69 | |
XM_007644818.1 | XP_007643008.1 | 30 - 31 | 4684 | 69 | ||
GenBank mRNA | JI877755.1 | - | 7 - 8 | 1615 | 69 | |
JP048856.1 | - | 7 - 8 | 1614 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |