Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003494905.2 | XP_003494953.1 | 3 - 4 | 1031 | 88 | |
GenBank mRNA | JI893687.1 | - | 3 - 4 | 1024 | 88 | |
JP041355.1 | - | 3 - 4 | 1024 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |