Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hunk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495576.2 | XP_003495624.1 | 2 - 3 | 389 | 107 | |
GenBank mRNA | JI876093.1 | - | 1 - 2 | 290 | 107 | |
JP054324.1 | - | 1 - 2 | 290 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Probe spans exons |