Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfsf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007609214.1 | XP_007607404.1 | 1 - 2 | 63 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis Probe spans exons |