Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcxd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007618573.1 | XP_007616763.1 | 3 - 4 | 1406 | 66 | |
GenBank mRNA | JI888802.1 | - | 1 - 2 | 241 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |