Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tceanc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495721.2 | XP_003495769.1 | 3 - 4 | 464 | 60 | |
XM_007643636.1 | XP_007641826.1 | 3 - 4 | 593 | 60 | ||
XM_007643642.1 | XP_007641832.1 | 3 - 4 | 359 | 60 | ||
GenBank mRNA | JI874062.1 | - | 3 - 4 | 455 | 60 | |
JP049262.1 | - | 3 - 4 | 455 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |