Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506542.1 | XP_003506590.1 | 3 - 4 | 257 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |