Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g4e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007622134.1 | XP_007620324.1 | 3 - 4 | 398 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |